Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lipase from Porcine Pancreas

Catalog No. p-4643283 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g
10 g

Catalyzes hydrolysis of esters in aqueous solutions and synthesis of esters in non-aqueous solutions

Lipases are a subclass of esterases. They also produce hydroxyl and carboxylic groups through the hydrolysis of ester linkages in poly(ethylene terephthalate).

  • Synonyms: Steapsin, Glycerol ester hydrolase, Tween hydrolase, Tributyrase, PPL, Triacylglycerol acylhydrolase, Triacylglycerol lipase
  • CAS Number: 9001-62-1
  • EC Number: 232-619-9
  • MDL Number: MFCD00131509
  • Format: Tan powder
  • Specific Activity: >12,000U/g solid
  • Extinction Coefficient: E1%280=13.3
  • Isoelectric point (pI): Lipase A=4.9 and Lipase B=5.0
  • Partly soluble in water and insoluble in alcohol

Used industrially for resolution of chiral compounds and transesterification production of biodiesel.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Chiral Compounds Resolution, Transesterification Production of Biodiesel
Name Lipase
Quantity 10 g
Source Porcine Pancreas
Storage Requirements 0°C
Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.