Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Liver S9 Homogenate, Suspended in Potassium Chloride

Catalog No. ICN50411 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN50411 2 mL
ICN50412 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN50411 Supplier MP Biomedicals™ Supplier No. 0850411
Add to Cart
Add to Cart

Post Mitochondrial Supernatant

  • Rat Liver S9
  • Aroclor 1254 Induced
  • Suspended in potassium chloride

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Ames mutagen assay, study of xenobiotic metabolism and drug interactions
Content And Storage -70°C
Source Male Rat, Sprague Dawley strain
Quantity 2 mL
Product Type Liver S9 Homogenate
Form Frozen
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.