Learn More
Description
Activity: >9000 units/mg protein. Unit Definition: One unit will cause a decrease in A450 of 0.001 per minute at pH 6.24 and 25°C using Micrococcus lysodeikticus as substrate.
- Lyophilized
- Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
- CAS: 9001-63-2
Specifications
Specifications
| Concentration | 2X |
| Color | White |
| pH | 11 |
| For Use With (Application) | Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen |
| Content And Storage | 0°C |
| Source | Chicken egg white |
| Quantity | 5 g |
| Product Type | Lysozyme |
| Molecular Weight (g/mol) | 14388 |
| Inhibitors | Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
| Show More |
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.