Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ M9 Minimal Medium Salts (Maniatis Formulation), Powdered

Catalog No. p-4474841 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
227 g
1000 g

Catalog No. MP113037012

Supplier: MP Biomedicals™ 113037012

Only null left
Add to Cart
Add to Cart

Used for preparing a defined synthetic medium for the growth of E. coli host

  • A commonly used base for preparing a defined synthetic medium for the growth of most E. coli host cells
  • Powder is reconstituted in sterile purified water, (or water with melted purified agar added)
  • Additional nutrients, such as glucose or other carbon source, casamino acids, vitamins, or antibiotics are added when the solution cools
  • Add additional nutrients before plates are poured or media is inoculated with bacteria
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified
Concentration 56.4 g/L of a 5X Solution
Content And Storage 15° to 30°C
Description A 56.4 g sample of M9 Minimal Salts was dissolved in an aqueous solution to 1 liter, the pH adjusted to 7.4 (if needed), and the solution autoclaved to make a sterile 5X concentrate.
Form Powder
Media Type M9 Minimal Medium
Product Type Microbiology Media
Sterility Sterile
Formulation 33.9 g Na2HPO4; 15 g KH2PO4; 2.5 g NaCl; 5 g NH4Cl
For Use With (Application) Preparing a Defined Synthetic Medium for The Growth of E. Coli Host
pH 6.8
Quantity 227 g
Shipping Condition Ambient
Solubility Soluble Aqueous Solution
Specimen Type Bacterial
Strain Designations E. coli
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.