Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Mouse Serum

Catalog No. p-2625437
Click to view available options
Quantity:
1 mL
5 mL
10 mL
Form:
Liquid
Lyophilized
Hemoglobin Concentration:
5.8 g/dL
9.65 mg/dL

Used for blocking agents and controls in immunoassays.

TRUSTED_SUSTAINABILITY

Specifications

Color Brown
Content And Storage -20°C
Species Mouse
Form Liquid
Product Type Mouse Serum
Quantity 1 mL
Sterilization Method 0.2 μm filtration
For Use With (Application) Blocking agents and controls in immunoassays
Hemoglobin Concentration 9.65 mg/dL
pH 7.4 to 8.1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.