Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Papain

Catalog No. ICN10256690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10256690 500 g
ICN10256680 100 g
ICN10256683 250 g
ICN10256691 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10256690 Supplier MP Biomedicals™ Supplier No. 0210256690
Add to Cart
Add to Cart

Used in the enzymatic synthesis of amino acids, peptides, and other molecules

Sulfhydryl protease from Carica papaya latex. (A second protease, chymopapain, and a lysozyme have also been isolated from this same source.)

  • Format: Powder
  • Grade: Technical
  • Molecular weight: 23,000
  • Specific Activity:>1750 USP Units/mg

TRUSTED_SUSTAINABILITY

Specifications

Color Off-White
pH 6 to 7
For Use With (Application) Enzymatic synthesis of Amino Acids, Peptides and other molecules
Content And Storage 4°C
Source Carica papaya latex
Quantity 500 g
Product Type Papain
Molecular Weight (g/mol) 23000
Inhibitors AEBSF, Antipain, Cystatin, E-64, leupeptin, PMSF, TLCK and TPCK
Form Powder
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.