Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Penicillin G Potassium Salt, MP Biomedicals™

Catalog No. MP219453610 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mU
10 mU
100 μm
25 mU

Catalog No. MP219453610

Supplier: MP Biomedicals, Inc 0219453610

Only null left
Add to Cart
Add to Cart

Penicillin G Potassium Salt Cell culture reagent, White Powder

  • It is used as a cell culture additive as an antibiotics.
  • Penicillin G potassium Salt is a salt of Penicillin G.
  • Penicillin G is a hydrophobic, β-lactam antibiotic produced by Penicillium spp.
  • Penicillin G is also known as benzylpenicillin and contains a 6’ phenylacetyl side chain.
  • In acid aqueous solutions, Penicillin is reported to decompose into penillic acid and penilloic acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-98-4
MDL Number MFCD00036193
InChI Key HTQQVOYPSGNVPA-HBRCYENSSA-M
Synonym Benzylpenicillin Potassium Salt
IUPAC Name potassium [3-(2-chloro-10H-phenothiazin-10-yl)propyl]dimethylamine (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [K+].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(Cl)C=C12.CC1(C)S[C@@H]2[C@H](NC(=O)CC3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Physical Form Powder
pH 5 to 7.5 (3% aq soln)
Melting Point 214°C to 217°C (literature)
Potency 1,500 - 1,700 u/mg
Color White
Quantity 10 mU
Validated Application Cell Culture
Molecular Formula C33H36ClKN4O4S2
Beilstein 3832841EC
Solubility Information Soluble in water(100mg/mL).
Molecular Weight (g/mol) 691.34
Grade Cell Culture
Show More Show Less
RTECSNumber XH9700000
storageNote1 Store at +4°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.