Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Pepsin
From porcine stomach mucosa
$177.00 - $1422.00
Specifications
For Use With (Application) | Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies |
---|---|
pH Range | 1 to 3.5 |
Sample Type | Porcine Stomach Mucosa |
Content And Storage | 0°C |
Form | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ICN19536705
|
MP Biomedicals™
0219536705 |
5 g |
Each for $659.00
|
|
|||||
ICN19536701
|
MP Biomedicals™
0219536701 |
1 g |
Each for $177.00
|
|
|||||
ICN19536710
|
MP Biomedicals™
0219536710 |
10 g |
Each for $1,422.00
|
|
|||||
Description
Purified by chromatography. Activity: 2000-2400U/mg protein. Unit Definition: One unit will produce a Δ A280 of 0.001 per minute at pH 2.0 and 37°C, measured as TCA-soluble products using hemoglobin as substrate. Lyophilized powder, essentially salt-free.
Specifications
Used to digest Proteins and is commonly used in the preparation of Fab fragments from Antibodies | |
Porcine Stomach Mucosa | |
Lyophilized |
1 to 3.5 | |
0°C | |
Pepsin |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title