Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pepstatin A

Catalog No. p-3179310 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
250 mg

Catalog No. ICN19536805

Supplier: MP Biomedicals™ 0219536805

Only null left
Add to Cart
Add to Cart

Synthetic inhibitor for pepsin, renin, cathepsin D, and other acid proteases.

Pepstatin A, [26305-03-3]; Isoval-Val-Val-Sta-Ala-Sta

C34H63N5O9, F.W. 685.9; Melting Point: 235-237°C

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enzyme Assay, Inhibition Assay, Molecular Biology, Protein Purification, Sample Preparation
Molecular Weight (g/mol) 685.91
Name Pepstatin A
Quantity 5 mg
Storage Requirements 0°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.