Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Protamine Sulfate

Catalog No. ICN19472901 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10,000 mg
100,000 mg
1000 mg
25000 mg
5000 mg

From salmon sperm

Cell Culture Reagent. Highly purified. Nitrogen: Approx. 22.5%. Hygroscopic white powder.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Room Temperature
Format Powder
For Use With (Application) Antiheparin activity, refining of vaccines, removal of DNA from Protein samples
Product Type Protamine Sulfate from Salmon Sperm
Quantity 1000 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.