Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RPM™ Kit

Catalog No. p-4696817 Shop All MP Biomedicals, Inc Products
Click to view available options
No. of Reactions:
120 Preps
300 Preps

For Nucleic Acid Isolation and Purification, Sample Lysis and Preparation

  • Kit is designed for rapid isolation and purification of double-stranded plasmid DNA from bacterial cultures. Theprocess yields pure plasmid DNA that is suitable for enzymatic manipulations including DNA sequencing, restriction enzymedigestion, in vitro transcription and PCR.
  • For research use only
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For rapid isolation, purification of double-stranded plasmid DNA from bacterial culture
Includes Pre-Lysis, Alkaline Lysis, Neutralizing, Glassmilk spin buffers, wash soln. concentrate
No. of Reactions 120 Preps
Product Type RPM Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.