Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Streptavidin
Click to view available options
Quantity:
1 mg
5 mg
Description
Has four binding sites for biotin, as does avidin. Both have been used extensively as probes in immunochemical systems, conjugated to antibodies, enzymes or fluorochromes.
- Format: Off-white clumpy powder
- Extinction Coefficient (E1%): E1% = 34 at 280nm (solvent: 0.1 M ammonium perchlorate, pH 8.0)(Lit.)
- Specific Activity: ∽10units/mg protein

Specifications
Specifications
Molecular Weight (g/mol) | 55000 |
Quantity | 1 mg |
Source | Streptomyces avidinii |
Content And Storage | 0°C |
Form | Lyophilized |
Product Type | Streptavidin from Streptomyces avidinii |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction