Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Terrific Broth II, Dehydrated Media

Catalog No. MP113046032 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP113046032 1000 g
MP113046022 454 g
MP113046042 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP113046032 Supplier MP Biomedicals™ Supplier No. 113046032
Add to Cart
Add to Cart

Rich medium for growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Grade Molecular Biology
Product Type Microbiology Media
Formulation Yeast extract, Tryptone broth, Sodium Chloride and Vitamins
For Use With (Application) For growth of Escherichia coli
Quantity 1000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.