Learn More
Description
Supplied in 50% glycerol/H2O.
Specifications
Specifications
| Color | Clear |
| For Use With (Application) | Used for site specific cleavage of recombinant fusion Proteins containing an accessible Thrombin recognition site for removal of affinity tags |
| Content And Storage | -20°C |
| Source | Bovine Plasma |
| Quantity | 200 μg |
| Product Type | alpha-Thrombin |
| Molecular Weight (g/mol) | 36700 |
| Form | Solution |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.