Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Trypsin Inhibitor, From Chicken Eggwhites
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
250 mg
0.5 g
1 g
5 g
Description
Trypsin Inhibitor, [9035-81-8]
- Essentially Salt-free
- Lyophilized
- 1 mg inhibits 1.0-3.0 mg of trypsin
- CAS # : 9035-81-8
- Activity: 1mg inhibits approxiately 2.5 to 3.0mg of trypsin.
1. Lineweaver and Murray, J. Biol. Chem., v. 171, 565 (1947)

Specifications
Specifications
Molecular Weight (g/mol) | 28500 |
Name | Trypsin Inhibitor |
Quantity | 5 g |
Source | Chicken Egg White |
Storage Requirements | 4°C |
Form | Lyophilized |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction