Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Tryptic Soybean Casein Digest Broth

Catalog No. ICN15715283 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15715283 250 g
ICN15715291 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15715283 Supplier MP Biomedicals™ Supplier No. 0215715283
Add to Cart
Add to Cart

Soybean casein digest broth

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Form Powder
Media Type Tryptic Soy Broth
Product Type Microbiology Media
Formulation Casein Peptone 17g/L, Sodium Chloride 5g/L, Dextrose 2.5g/L, Soy Peptone 3g/L, Dipotassium Phosphate 2.5gm/L
For Use With (Application) Isolation and cultivation of non-fastidious organisms
Quantity 250 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.