Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP EcoChrom Aluminum Oxide, Acid Technical A , MP Biomedicals, LLC

Catalog No. MP0207896 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 kg

MP EcoChrom Aluminum Oxide, Acid Technical A is a white to light pink free flowing powder that is ideal for quality control.

  • Ideal for the isolation of alkaloids, glycosides, active substances from plants, vitamins, hormones, and antibiotics
  • Dehydration of organic solvents
  • Purification of enzymes
  • Removal of terpenes, peroxides, and pyrogens from injectable solutions and infusions
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point ∼2980°C
Melting Point ∼2072°C
Color Pink to White
Quantity 100 kg
Density 3.94 g/cm³at 20 °C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.