Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP68 Polyclonal Antibody
MP68 Polyclonal Antibody
Supplier: Thermo Scientific PA563868
Description
MP68 Polyclonal Antibody for IHC

Specifications
MP68 | |
Polyclonal | |
Unconjugated | |
Atp5mpl | |
2010107E04Rik; 6.8 kDa mitochondrial proteolipid; 6.8 kDa mitochondrial proteolipid protein; 9430003J03Rik; ATP synthase membrane subunit 6.8PL; ATP synthase subunit ATP5MPL, mitochondrial; Atp5mpl; AU043134; AV124504; C14orf2; chromosome 14 open reading frame 2; MLQ; Mp68; PLPM; PRO1574; RIKEN cDNA 2010107E04 gene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P56378 | |
Atp5mpl | |
Recombinant protein corresponding to Human MP68. Recombinant protein control fragment (Product #RP-100879). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction