Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MP68 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112399
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112399 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112399 Supplier Invitrogen™ Supplier No. PA5112399
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MP68 Polyclonal Antibody for Western Blot, ELISA

MP68, also known as PLPM or C14orf2 (chromosome 14 open reading frame 2), is a 58 amino acid mitochondrial protein that belongs to the small mitochondrial proteolipid family. The gene encoding MP68 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MP68
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Atp5mpl
Gene Accession No. P56378
Gene Alias 2010107E04Rik; 6.8 kDa mitochondrial proteolipid; 6.8 kDa mitochondrial proteolipid protein; 9430003J03Rik; ATP synthase membrane subunit 6.8PL; ATP synthase subunit ATP5MPL, mitochondrial; Atp5mpl; AU043134; AV124504; C14orf2; chromosome 14 open reading frame 2; MLQ; Mp68; PLPM; PRO1574; RIKEN cDNA 2010107E04 gene
Gene Symbols Atp5mpl
Host Species Rabbit
Immunogen Recombinant Human 6.8 kDa mitochondrial proteolipid protein (1-58AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9556
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.