Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma mPAGE™ 4-12% Bis-Tris Precast Gel

Catalog No. 11580601 Shop All MilliporeSigma Supelco Products
Click to view available options
Wells:
10-well
12-well
15-well

Precast gel for research use only

  • mPAGE 4-12% Bis-Tris Precast Gel, 10x8 cm
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
TRUSTED_SUSTAINABILITY

Specifications

Buffer MES or MOPS Running Buffer;
Concentration 4 to 12% Bis to Tris
For Use With (Equipment) Mini Gel Systems
Gel Percentage 4 to 12%
Gel Thickness 1 mm
Gel Type Bis-Tris
Product Type Bis-Tris precast gels
Sample Loading Volume 80 μL
Separation of Protein
Wells 10-well
Quantity 10 Pk.
Length (Metric) 10 cm
Width (Metric) 8 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.