Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MPC 6827 hydrochloride
SDP

Catalog No. 523150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-315-0 50 mg
52-311-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-315-0 Supplier Tocris Bioscience™ Supplier No. 5231/50

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of microtubule polymerization; antimitotic and antitumor

MPC 6827 hydrochloride is a potent inhibitor of microtubule formation (IC50 = 1.5 - 3.4 nM); inhibits polymerization of tubulin in vitro and disrupts microtubule formation in several cancer cell lines. Inhibits tumor growth in vitro and in vivo; exhibits pro-apoptotic characteristics. Brain penetrant.

Specifications

Purity >98%
Quantity 50 mg
Product Type MPC 6827 hydrochloride
Molecular Weight (g/mol) 315.8
Inhibitors MPC 6827 hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.