Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPDU1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP18457025UL
Description
MPDU1 Polyclonal specifically detects MPDU1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MPDU1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
CDGIF, FLJ14836, HBeAg-binding protein 2 binding protein A, Lec35, mannose-P-dolichol utilization defect 1, mannose-P-dolichol utilization defect 1 protein, My008, PP3958, PQLC5, SL15HBEBP2BPA, Suppressor of Lec15 and Lec35 glycosylation mutation homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
9526 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MPDU1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:AEADGPLKRLLVPILLPEKCYDQLFVQWDLLHVPCLKILLSK | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction