Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPDU1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546790 Supplier Invitrogen™ Supplier No. PA546790
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LARIFTSIQE TGDPLMAGTF VVSSLCNGLI AAQLLFYWNA KPPHKQKKAQ.

This gene encodes an endoplasmic reticulum membrane protein that is required for utilization of the mannose donor mannose-P-dolichol in the synthesis of lipid-linked oligosaccharides and glycosylphosphatidylinositols. Mutations in this gene result in congenital disorder of glycosylation type If. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPDU1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MPDU1
Gene Accession No. O75352
Gene Alias CDGIF; HBeAg-binding protein 2 binding protein A; HBEBP2BPA; Lec35; mannose-P-dolichol utilization defect 1; mannose-P-dolichol utilization defect 1 protein; MPDU1; My008; PP3958; PQLC5; SL15; Supl15h; Suppressor of Lec15 and Lec35 glycosylation mutation homolog; suppressor of Lec15 homolog (C.griseus)
Gene Symbols MPDU1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human MPDU1 (aa 198-247).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9526
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.