Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579691
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579691 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579691 Supplier Invitrogen™ Supplier No. PA579691
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Liver Tissue, HELA whole cell, JURKAT whole cell.

MPG functions in the hydrolysis of the deoxyribose N-glycosidic bond to excise 3-methyladenine, and 7-methylguanine from the damaged DNA polymer formed by alkylation lesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPG
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene MPG
Gene Accession No. P23571, P29372, Q04841
Gene Alias 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p; 3-alkyladenine DNA glycosylase; 3-methyladenine DNA glycosidase; 3-methyladenine DNA glycosylase; 9830006D05; AAG; ADPG; AI326268; alkylpurine-DNA-N-glycosylase; ANPG; APNG; CRA36.1; CRA36.1 (3-methyl-adenine DNA glycosylase); DNA-3-methyladenine glycosylase; MDG; Mid1; MPG; N-methylpurine DNA glycosylase; N-methylpurine-DNA glycocylase; N-methylpurine-DNA glycosylase; N-methylpurine-DNA glycosylase, MPG; PIG11; PIG16; proliferation-inducing protein 11; proliferation-inducing protein 16
Gene Symbols MPG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human MPG (228-243aa NKSFDQRDLAQDEAVW).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24561, 268395, 4350
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.