Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPG Polyclonal Antibody, Invitrogen™

Catalog No. PIPA596313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596313 Supplier Thermo Scientific Supplier No. PA596313

Rabbit Polyclonal Antibody

Immunogen sequence: MPARSGAQFC RRMGQKKQRP ARAGQPHSSS DAAQAPAEQP HSSSDAAQAP CPRERCLGPP TTPGPYRSIY FSSPKGHLTR LGLEFFDQPA VPLARAFLGQ VLVRRLPNGT ELRGRIVETE AYLGPEDEAA HSRGGRQTPR NRGMFMKPGT LYVYIIYGMY FCMNISSQGD GACVLLRALE PLEGLETMRQ LRSTLRKGTA SRVLKDRELC SGPSKLCQAL AINKSFDQRD LAQDEAVWLE RGPLEPSEPA VVAAARVGVG HAGEWARKPL RFYVRGSPWV SVVDRVAEQD TQA; Positive Samples: MCF7, K-562, U-937, Mouse liver; Cellular Location: Cytoplasm, Mitochondrion matrix, Nucleus, mitochondrion nucleoid

MPG functions in the hydrolysis of the deoxyribose N-glycosidic bond to excise 3-methyladenine, and 7-methylguanine from the damaged DNA polymer formed by alkylation lesions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPG
Applications Immunocytochemistry, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide, pH 7.3
Gene MPG
Gene Accession No. P29372, Q04841
Gene Alias 3' end of the Mid1 gene, localized 68 kb upstream the humanzeta globin gene on 16p, 3-alkyladenine DNA glycosylase, 3-methyladenine DNA glycosidase, 3-methyladenine DNA glycosylase, 9830006D05, AAG, ADPG, AI326268, alkylpurine-DNA-N-glycosylase, ANPG, APNG, CRA36.1, CRA36.1 (3-methyl-adenine DNA glycosylase), DNA-3-methyladenine glycosylase, MDG, Mid1, MPG, N-methylpurine DNA glycosylase, N-methylpurine-DNA glycocylase, N-methylpurine-DNA glycosylase, N-methylpurine-DNA glycosylase, MPG, PIG11, PIG16, proliferation-inducing protein 11, proliferation-inducing protein 16
Gene Symbols MPG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-293 of human MPG (NP_0010150521)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268395, 4350
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.