Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPHOSPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599654
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599654 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599654 Supplier Invitrogen™ Supplier No. PA599654
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MPP1.

KIF20B is a plus-end-directed motor enzyme that is required for completion of cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPHOSPH1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIF20B
Gene Accession No. Q80WE4, Q96Q89
Gene Alias 33cex; B130024C23; C330014J10Rik; Cancer/testis antigen 90; CT90; Kif20b; Kinesin family member 20B; kinesin-like protein KIF20B; Kinesin-related motor interacting with PIN1; KRMP1; magoo; mitotic kinesin-like protein; mitotic kinesin-related protein; M-phase phosphoprotein 1; MPHOSPH1; MPP1; MPP-1; N-6 kinesin
Gene Symbols KIF20B
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIF20B(Accession Q96Q89), corresponding to amino acid residues M1-E49.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240641, 9585
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.