Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPHOSPH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA539253
Description

Specifications
MPHOSPH1 | |
Polyclonal | |
Unconjugated | |
KIF20B | |
KRMP1, MPHOSPH1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
240641, 9585 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96Q89, Q80WE4 | |
KIF20B | |
A synthetic peptide derived from the C-terminal region of human KIF20B | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction