Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MPHOSPH6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109972
Description
Immunogen sequence: MAAERKTRLS KNLLRMKFMQ RGLDSETKKQ LEEEEKKIIS EEHWYLDLPE LKEKESFIIE EQSFLLCEDL LYGRMSFRGF NPEVEKLMLQ MNAKHKAEEV EDETVELDVS DEEMARRYET LVGTIGKKFA RKRDHANYEE DENGDITPIK AKKMFLKPQD.
MPHOSPH6 (M Phase Phosphoprotein 6) is a Protein Coding gene. Among its related pathways areGene ExpressionandDeadenylation dependent mRNA decay. Gene Ontology (GO) annotations related to this gene includeRNA binding.
Specifications
| MPHOSPH6 | |
| Polyclonal | |
| Unconjugated | |
| MPHOSPH6 | |
| 1110001M01Rik; AA536809; C86426; M phase phosphoprotein 6; M-phase phosphoprotein 6; Mphosph6; MPP; MPP6; MPP-6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10200, 68533, 686999 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.10 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q99547, Q9D1Q1 | |
| MPHOSPH6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human MPHOSPH6 (NP_005783.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction