Learn More
MPI Mouse anti-Human, Clone: OTI4G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504772

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.Specifications
MPI | |
Monoclonal | |
Unconjugated | |
MPI | |
CDG1B, PMI, PMI1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4351 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI4G7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P34949 | |
MPI | |
Full length human recombit protein of human MPI produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.