Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582287 Supplier Invitrogen™ Supplier No. PA582287
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSNSEVARLL ASSDPLAQIA EDKPYAELWM GTHPRGDAKI LDNRISQKTL SQWIAENQDS LGSKVKDTFN GNLPFLFKVL SVETPLSIQA HPNKELAEKL HLQAPQHYPD ANHKPEMAIA LTPFQGLCG Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 91%.

Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPI
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MPI
Gene Accession No. P34949
Gene Alias 1110002E17Rik; AI315153; CDG1B; mannose phosphate isomerase; mannose phosphate isomerase (mapped); mannose phosphate isomerase 1; mannose-6-phosphate isomerase; MPI; Mpi1; Mpi-1; Phosphohexomutase; phosphomannose isomerase; phosphomannose isomerase 1; PMI; PMI1
Gene Symbols MPI
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MPI. Recombinant protein control fragment (Product #RP-89702).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4351
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.