Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596635 Supplier Invitrogen™ Supplier No. PA596635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAPRVFPLS CAVQQYAWGK MGSNSEVARL LASSDPLAQI AEDKPYAELW MGTHPRGDAK ILDNRISQKT LSQWIAENQD SLGSKVKDTF NGNLPFLFKV LSVETPLSIQ AHPNKELAEK LHLQAPQHYP DANHKPEMAI ALTPFQGLCG FRPVEEIVTF LKKVPEFQFL IGDEAATHLK QTMSHDSQAV ASSLQSCFSH LMKSEKKVVV EQLNLLVKRI SQQAAAGNNM EDIFGELLLQ LHQQYPGDIG CFAIYFLNLL TLKPGEAMFL EANVPHAYLK GDCVECMACS DNTVRAGLTP KFIDVPTLCE MLSYTPSSSK DRLFLPTRSQ EDPYLSIYDP PVPDFTIMKT EVPGSVTEYK VLALDSASIL LMVQGTVIAS TPTTQTPIPL QRGGVLFIGA NESVSLKLTE PKDLLIFRAC CLL; Positive Samples: Jurkat, HepG2, HeLa, SW620, Mouse heart; Cellular Location: Cytoplasm.

Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPI
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MPI
Gene Accession No. P34949, Q68FX1, Q924M7
Gene Alias 1110002E17Rik; AI315153; CDG1B; mannose phosphate isomerase; mannose phosphate isomerase (mapped); mannose phosphate isomerase 1; mannose-6-phosphate isomerase; MPI; Mpi1; Mpi-1; Phosphohexomutase; phosphomannose isomerase; phosphomannose isomerase 1; PMI; PMI1
Gene Symbols MPI
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-423 of human MPI (NP_0024261).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110119, 300741, 4351
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.