Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPI Polyclonal Antibody

Catalog No. PIPA5121767
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121767 20 μL
PIPA5119270 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121767 Supplier Invitrogen™ Supplier No. PA5121767
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MPI Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Phosphomannose isomerase catalyzes the interconversion of fructose-6-phosphate and mannose-6-phosphate and plays a critical role in maintaining the supply of D-mannose derivatives, which are required for most glycosylation reactions. Mutations in the MPI gene were found in patients with carbohydrate-deficient glycoprotein syndrome, type Ib.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPI
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene MPI
Gene Accession No. P34949
Gene Alias 1110002E17Rik; AI315153; CDG1B; mannose phosphate isomerase; mannose phosphate isomerase (mapped); mannose phosphate isomerase 1; mannose-6-phosphate isomerase; MPI; Mpi1; Mpi-1; Phosphohexomutase; phosphomannose isomerase; phosphomannose isomerase 1; PMI; PMI1
Gene Symbols MPI
Host Species Rabbit
Immunogen Recombinant protein Mannose Phosphate Isomerase. The antigen corresponds to amino acid range 2-423 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100154551, 4351
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.