Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MPK-3 (DUSP6), Mouse, Unlabeled, Clone: 5H6.1,

Catalog No. MABS1726 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABS1726 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1726 Supplier MilliporeSigma™ Supplier No. MABS1726
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Dual specificity protein phosphatase 6 (EC 3.1.3.16; EC 3.1.3.48; UniProt Q16828; also known as Dual specificity protein phosphatase PYST1, MAP kinase phosphatase 3, Mitogen-activated protein kinase phosphatase 3, MKP-3) is encoded by the DUSP6 (also known as HH19, MKP3, PYST1) gene (Gene ID 1848) in human. MPK-3 (DUSP6) belongs to the family of dual-specificity phosphatases (DUSPs) that mediate dephosphorylation of MAPKs on threonine/serine and tyrosine residues. DUSP6 is upregulated in HER2+ carcinomas and is among several DUSPs thought to be involved in breast cancer metastasis. DUSP1, DUSP4, and DUSP6 are induced during epithelial-to-mesenchymal transition (EMT) and are involved in forming and maintaining breast cancer stem cells (CSCs), DUSP1 knockdown reduces CSC formation, while DUSP4 or DUSP6 knockdown enhances CSC formation. PKC CK2alpha is reported to phosphorylate DUSP6 and increase its dephosphatase activity. ERK1/2, on the other hand, can phosphorylate DUSP6 to promote its proteosomal degradation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPK-3 (DUSP6)
Applications Western Blot
Classification Monoclonal
Clone 5H6.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2bκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene DUSP6, HH19, MKP3, PYST1
Gene Accession No. Q16828
Host Species Mouse
Immunogen GST-tagged recombinant human MPK-3 (DUSP6) rhodanese domain fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001937
Test Specificity Clone 5H6.1 targets an epitope within the rhodanese domain present in both spliced isoforms of human MPK-3 (DUSP6) reported by UniProt (Q16828).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.