Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPLKIP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116262
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116262 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116262 Supplier Invitrogen™ Supplier No. PA5116262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MPLKIP Polyclonal Antibody for Western Blot

May play a role in maintenance of cell cycle integrity by regulating mitosis or cytokinesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPLKIP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene MPLKIP
Gene Accession No. Q8TAP9
Gene Alias ABHS; C7orf11; M-phase specific PLK1 interacting protein; M-phase-specific PLK1-interacting protein; MPLKIP; ORF20; Russell-Silver syndrome region; TTD non-photosensitive 1 protein; TTD4; TTDN1
Gene Symbols MPLKIP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 151-177 amino acids from the C-terminal region of human TTDN1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136647
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.