Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mPlum Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP311833UV
Description
mPlum Polyclonal antibody specifically detects mPlum in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
| mPlum | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| DyLight 350 | |
| Fluorescent protein plum, mPlum Fluorescent Protein, Red Fluorescent Protein, RFP | |
| mPlum from Discosoma sp. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction