Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mPlum Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP311833C
Description
mPlum Polyclonal antibody specifically detects mPlum in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, ImmunofluorescenceSpecifications
mPlum | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Non-species specific | |
Purified |
Western Blot, Flow Cytometry, Immunocytochemistry | |
DyLight 650 | |
Fluorescent protein plum, mPlum Fluorescent Protein, Red Fluorescent Protein, RFP | |
mPlum from Discosoma sp. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction