Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPND Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP17926320UL

 View more versions of this product

Catalog No. NBP17926320


Description

Description

MPND Polyclonal specifically detects MPND in Human samples. It is validated for Western Blot.
Specifications

Specifications

MPND
Polyclonal
Western Blot 1:1000
NP_116257
MPND
Synthetic peptide directed towards the N terminal of human MPNDThe immunogen for this antibody is MPND. Peptide sequence MAAPEPLSPAGGAGEEAPEEDEDEAEAEDPERPNAGAGGGRSGGGGSSVS.
20 μL
Primary
Human
Purified
Western Blot
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
EC 3.4, FLJ14981, MPN domain containing, MPN domain-containing protein
Rabbit
Protein A purified
RUO
84954
Store at -20C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.