Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MPND Monoclonal Antibody (1C12)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA522808
Description
Peptide Sequence: ALLEPGAGVL SIYYLGKKFL GDLQPDGRIM WQETGQTFNS PSAWATHCKK LVNPAKKSGC GWASVKYKGQ KLDKYKATWL RLHQLHTPAT AADESPAS.
Probable protease. Acts as a sensor of N(6)-methyladenosine methylation on DNA (m6A): recognizes and binds m6A DNA, leading to its degradation (PubMed:30982744). [UniProt].
Specifications
| MPND | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8N594 | |
| MPND | |
| MPND (NP_116257, 84 a.a. ~ 181 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| 1C12 | |
| Unconjugated | |
| MPND | |
| E130307M08Rik; FLJ14981; MPN domain containing; MPN domain-containing protein; Mpnd | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84954 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction