Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPND Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734348 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4348 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4348 Supplier Proteintech Group Inc Supplier No. 244201AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Probable protease.

Specifications

Antigen MPND
Applications Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MPND
Gene Accession No. Q3TV65, Q8N594
Gene Alias MPN domain containing, MPN domain containing protein, MPND
Gene Symbols MPND
Host Species Rabbit
Immunogen MPND Fusion Protein Ag19531
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 68047, 84954
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.