Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPND Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$480.74
Specifications
| Antigen | MPND |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179262
![]() |
Novus Biologicals
NBP179262 |
100 μL |
Each for $480.74
|
|
|||||
NBP17926220
![]() |
Novus Biologicals
NBP17926220UL |
20 μL | N/A | N/A | N/A | ||||
Description
MPND Polyclonal specifically detects MPND in Human samples. It is validated for Western Blot.Specifications
| MPND | |
| Polyclonal | |
| Rabbit | |
| NP_116257 | |
| 84954 | |
| Synthetic peptide directed towards the middle region of human MPNDThe immunogen for this antibody is MPND. Peptide sequence PASEGEEEELLMEEEEEDVLAGVSAEDKSRRPLGKSPSEPAHPEATTPGK. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| EC 3.4, FLJ14981, MPN domain containing, MPN domain-containing protein | |
| MPND | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title