Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPO Mouse anti-Human, Clone: 4C11F6, Proteintech
SDP

Catalog No. 501736611 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6611 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6611 Supplier Proteintech Group Inc Supplier No. 661771IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myeloperoxidase (MPO) is a hemoprotein that is abundantly expressed in neutrophils and secreted during their activation. Native Myeloperoxidase is represented as a covalently bound tetrameric complex of two glycosylated alpha chains with a molecular weight range of 59 - 64 kDa, and two unglycosylated beta chains (MW approximately 14 kDa) with total MW approximately 150 kDa and a theoretical pI of 9.2. Traditionally, myeloperoxidase was considered as a main target of anti-neutrophil cytoplasm antibodies (ANCA), the serological markers for certain systemic vasculities such as periarteriitis nodosa, microscopic polyarteriitis and pulmonary eosinophilic granulomatosis (Churg-Strauss syndrome). Low to moderate anti-myeloperoxidase autoantibody levels are also reported in rheumatoid arthritis. Recently, it was shown that myeloperoxidase participates in the initiation and progression of cardiovascular disease. Myeloperoxidase possesses potent proinflammatory properties and may contribute directly to tissue injury. Now Myeloperoxidase is under consideration as one of the most promising cardiac markers. Myeloperoxidase is part of the host defense system of polymorphonuclear leukocytes and myeloperoxidase is responsible for microbicidal activity against a wide range of organisms. In the stimulated PMN (polymorphonuclear leukocytes), MPO catalyzes the production of hypohalous acids, primarily hypochlorous acid in physiologic situations, and other toxic intermediates that greatly enhance PMN microbicidal activity.

Specifications

Antigen MPO
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4C11F6
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MPO
Gene Accession No. P05164
Gene Alias MPO, myeloperoxidase
Gene Symbols MPO
Host Species Mouse
Immunogen MPO Fusion Protein Ag17564
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4353
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.