Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ MPO (Mouse) ELISA Kit

Catalog No. p-4747590
Click to view available options
Conjugate:
Biotin
HRP (Horseradish Peroxidase)
Sample Type:
Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Cell Culture Supernatant, Plasma

Includes

  • 96-well plate precoated with anti-mouse MPO antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse MPO standard, 10ng x 2
  • Biotinylated anti-mouse MPO antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively mouse Mpo. Sandwich enzyme immunoassay.

Mpo (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Assay Sensitivity <10 pg/mL
Calibration Range 156 to 10000pg/mL
Content And Storage Store the kit at -20°C
Description Mpo (Mouse) is a sandwich enzyme immunoassay for the quantitative measurement of mouse Mpo.
Conjugate Biotin
Cross Reactivity Mouse
Detection Method Colorimetric
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Homogenates
Specificity Natural and recombinant mouse MPO
Sufficient For 96
For Use With (Application) Quantification
Kit Contents 96-well plate precoated with anti- mouse MPO antibody (96 (8 x 12) wells), Lyophilized recombinant mouse MPO standard (10ng x 2), Biotinylated anti- mouse MPO antibody (dilution 1:100): 13μL, Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100): 130μL, Sample diluent buffer (30mL), Antibody diluent buffer (12mL), ABC diluent buffer (12mL), TMB color developing agent (10mL), TMB stop solution (10mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Volume 100 μL
Detection Limit <10 pg/mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.