Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MPP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | MPP1 |
---|---|
Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
MPP1 Polyclonal antibody specifically detects MPP1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MPP1 | |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, pH 7.2, 40% glycerol | |
4354 | |
IgG | |
Affinity purified |
Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
55 kDa erythrocyte membrane protein, AAG12, EC 2.7.11, Membrane protein, palmitoylated 1, membrane protein, palmitoylated 1, 55kDa, MRG1, palmitoylated erythrocyte membrane protein, PEMPpalmitoylated 1 (55kD) | |
This antibody was developed against Recombinant Protein corresponding to amino acids: MTLKASEGESGGSMHTALSDLYLEHLLQKRSRPEAVSHPLNTVTEDMYTNGSPAPGSPAQVKGQEVRKVRLIQFEKVTE | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title