Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MPP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557135
Description
Immunogen sequence: DDDLQENEDN KQHKESLKRV TFALPDDAET EDTGVLNVKK NSDEVKSSFE KRQEKMNEKI ASLEKELLEK KPWQLQGEVT AQKRPENSLL EETLHFDHAV R Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
MPP10 (M-phase phosphoprotein 10), also known as MPHOSPH10, is a 681 amino acid protein that localizes to the fibrillar region of the nucleolus. Phosphorylated in the M (mitotic) phase of the cell cycle, MPP10 is a component of the 60-80S U3 small nucleolar ribonucleoprotein (U3 snoRNP) complex and is required for the early cleavages during pre-18S ribosomal RNA processing. MPP10 forms a heterotrimeric complex with IMP-3 and IMP-4, which may be required for its association with nucleolar components. The gene encoding MPP10 is localized to human chromosome 2, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia.
Specifications
MPP10 | |
Polyclonal | |
Unconjugated | |
MPHOSPH10 | |
2810453H10Rik; 5730405D16Rik; AI326008; CT90; M phase phosphoprotein 10; M-phase phosphoprotein 10; M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein); Mphosph10; MPP10; MPP10P; PPP1R106; protein phosphatase 1, regulatory subunit 106; U3 small nucleolar ribonucleoprotein; U3 small nucleolar ribonucleoprotein protein MPP10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10199 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00566 | |
MPHOSPH10 | |
Recombinant protein corresponding to Human MPP10. Recombinant protein control fragment (Product #RP-96066). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction