Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521526
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521526 Supplier Invitrogen™ Supplier No. PA521526
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549, HeLa, HCT116, mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (82%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The MAGUK (membrane-associated guanylate kinase homologs) family of proteins contain multiple protein-binding domains and are involved in cell junction organization, tumor suppression and signaling. The MAGUK family is divided into four subfamilies: DLG-like, ZO1-like, p55-like and LIN2-like. MPP2 (membrane protein, palmitoylated 2), also known as MAGUK p55 subfamily member 2, discs large homolog 2 or DLG2, is a 576 amino acid protein belonging to the MAGUK family that exists as three alternatively spliced isoforms. MPP2 contains one guanylate kinase-like domain, a PDZ (DHR) domain, two L27 domains and a single SH3 domain. The gene encoding MPP2 maps to the same segment of human chromosome 17 as MPP3, with whom MMP2 likely shares similar function and common structural organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPP2
Applications Western Blot
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Mpp2
Gene Accession No. Q14168, Q9WV34
Gene Alias D11Bwg0652e; discs large homolog 2; discs large, homolog 2; Dlg2; Dlgh2; MAGUK p55 subfamily member 2; membrane palmitoylated protein 2; membrane protein, palmitoylated 2 (MAGUK p55 subfamily member 2); Mpp2; p55 subfamily member of MAGUK family; palmitoylated membrane protein 2; Pals4; Protein MPP2; rCG_34691
Gene Symbols Mpp2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MPP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4355, 50997
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.