Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MPP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570670
Description
This target displays homology in the following species: Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 86%; Zebrafish: 85%.
MPP4 is a protein coding gene.
Specifications
MPP4 | |
Polyclonal | |
Unconjugated | |
MPP4 | |
ALS2CR5; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 5 protein; discs large homolog 6; discs, large homolog 6; DLG6; MAGUK p55 subfamily member 4; mDLG6; membrane palmitoylated protein 4; membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4); Mpp4; rDLG6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
58538 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96JB8 | |
MPP4 | |
synthetic peptide directed towards the middle region of human MPP4 (aa 434-483). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction