Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MPP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584341
Description
Immunogen sequence: QALLKIYDCL QEFKEKKLVP ATPHAQVLSY EVVELLRETP TSPEIQELRQ MLQAPHFKAL LSAHDTIAQK D.
MPP4 is a protein coding gene.
Specifications
MPP4 | |
Polyclonal | |
Unconjugated | |
MPP4 | |
ALS2CR5; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 5 protein; discs large homolog 6; discs, large homolog 6; DLG6; MAGUK p55 subfamily member 4; mDLG6; membrane palmitoylated protein 4; membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4); Mpp4; rDLG6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
58538 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JB8 | |
MPP4 | |
Recombinant protein corresponding to Human MPP4. Recombinant protein control fragment (Product #RP-108422). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction