Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MPP4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983716
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3716 Supplier Bioss Supplier No. BS15379R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MPP4 is a protein coding gene.

Specifications

Antigen MPP4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MPP4
Gene Accession No. Q96JB8
Gene Alias ALS2CR5; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 5 protein; Discs large homolog 6; discs, large homolog 6; DLG6; MAGUK p55 subfamily member 4; mDLG6; membrane palmitoylated protein 4; membrane protein, palmitoylated 4 (MAGUK p55 subfamily member 4); MPP4; rDLG6
Gene Symbols MPP4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MPP4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58538
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.