Learn More
Invitrogen™ MPP7 Polyclonal Antibody

Description
Sequence homology: Cow: 100%; Dog: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Specifications
Specifications
| Antigen | MPP7 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 |
| Gene | MPP7 |
| Gene Accession No. | Q5T2T1 |
| Gene Alias | 1110068J02Rik; 2810038M04Rik; 5430426E14Rik; AI415104; Gm955; MAGUK p55 subfamily member 7; membrane palmitoylated protein 7; membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7); MPP7 |
| Gene Symbols | MPP7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.