Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MPP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583380
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583380 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583380 Supplier Invitrogen™ Supplier No. PA583380
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QERRLALRRP EILVQPLKVS NRKSSGFRRS FRLSRKDKKT NKSMYECKKS DQYDTADVPT YEEVT Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 95%.

Acts as an important adapter that promotes epithelial cell polarity and tight junction formation via its interaction with DLG1. Involved in the assembly of protein complexes at sites of cell-cell contact.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MPP7
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MPP7
Gene Accession No. Q5T2T1
Gene Alias 1110068J02Rik; 2810038M04Rik; 5430426E14Rik; AI415104; Gm955; MAGUK p55 subfamily member 7; membrane palmitoylated protein 7; membrane protein, palmitoylated 7 (MAGUK p55 subfamily member 7); MPP7
Gene Symbols MPP7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MPP7. Recombinant protein control fragment (Product #RP-105936).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.